Learn More
Corning™ 96-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate
Shop All Corning Products
Click to view available options
Color:
Black, Clear Bottom
Unit Size:
Pack of 100
Description
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Culture Area | 0.32 cm2 |
| Array | 8 x 12 |
| Color | Black, Clear Bottom |
| Material | Polystyrene |
| No. of Wells | 96 |
| Volume (Metric) Working | 75 to 200 μL |
| Sterility | Sterile |
| Surface Treatment | Poly-D-Lysine |
| Show More |