Learn More
Corning™ DeckWorks™ Low-Binding Hinged Rack Pipet Tips, Nonsterile
Product Code. 12999577 Shop All Corning Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 200 μL
1 to 300 μL
Unit Size:
Pack of 3840
Pack of 960
Product Code. 12999577
Brand: Corning™ 4149
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for quantitative assays in which sample binding can significantly alter results.
Corning DeckWorks low binding tips are available in 10 μL, 200 μL, and 300 μL sizes. Proprietary low binding technology reduces surface tension on the interior wall of the tip. Leaching and/or subsequent sample degradation associated with silicone-based tips are eliminated. DeckWorks low binding tips are ideal for quantitative assays in which sample binding can significantly alter results. Specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins and/or peptides.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 1 to 300 μL
Sterility Non-sterile
Color Natural
For Use With (Application) Designed for quantitative assays in which sample binding can significantly alter results
Format Hinged Rack
Tip Style Low Retention, Round
Packaging Hinged rack
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line DeckWorks
Material Polypropylene