Learn More
Corning™ Costar™ Transtar™-96 96-Well Liquid Transfer System Accessories: Reservoir Liners
Shop All Corning Products
Click to view available options
Type:
Open Reservoir Liner
Reservoir
Unit Size:
Pack of 100
Description
For use with 96-well plates
These are sterile replacement open reservoir liners for the Transtar-96™ Well Liquid Transfer system.
Spécification
Spécification
| Color | White |
| Material | Polystyrene |
| Sterility | Sterile |
| Type | Reservoir |
| For Use With (Application) | Ideal for repetitively filling multi-channel pipettors |
| No. per Case | 100/Cs. |
| No. per Pack | 1/Pk. |
| Channels | 8 |