Learn More
JULABO CORIO CP-1200F Refrigerated/Heating Circulator with Natural Refrigerants
Refrigerated circulators from the CORIO CP range are suitable for applications with a temperature range up to +200°C. The enhanced pump performance ensures they are suitable for easy temperature control tasks in combination with external applications.
Brand: JULABO 9013717.N1
This item is not returnable.
View return policy
Description
- The optimized design of the bath tank and cooling coil makes the bath more spacious and user-friendly for the operator.
- Ventilation slots and connections are located on the front and rear, allowing several units to be placed next to each other in order to save space.
- Models for internal and external applications
- Bright, white, easy to read display
- Very quiet
- Powerful and infinitely adjustable pressure pump
- USB connection
- RS232 interface for online communication
- Bath lid and drain tap included
- Removable ventilation grid
- Refrigeration unit without side vents
- Class III (FL) according to DIN 12876-1
- Easy switching between internal and external circulation
- Integrated pump connection M16×1
- Space-saving cooling coil design yields more usable space in the bath tank
- Early warning system for low liquid level
Specifications
5 to 7.5 L | |
1.24kW at 20°C | |
Immersion Pump | |
Circulator |
RS232,USB | |
8 to 27 L/min. | |
-50°C to +200°C | |
200/230 V 50/60 Hz |