missing translation for 'onlineSavingsMsg'
Learn More

Copper iodate, anhydrous, 95%, Thermo Scientific Chemicals

€ 80.50 - € 1376.85

Chemical Identifiers

CAS 13454-89-2
Molecular Formula CuI2O6
Molecular Weight (g/mol) 413.35
MDL Number MFCD00016055
InChI Key LLVVIWYEOKVOFV-UHFFFAOYSA-L
Synonym copper iodate, copper 2+ diiodate, cupric iodate, copper iodate, anhydrous, cu.2io3
PubChem CID 9801829
IUPAC Name copper;diiodate
SMILES [Cu++].[O-][I](=O)=O.[O-][I](=O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11463780
View Documents
Thermo Scientific Alfa Aesar
089459.04
2 g
€ 80.50
2g
Please to purchase this item. Need a web account? Register with us today!
11453780
View Documents
Thermo Scientific Alfa Aesar
089459.09
10 g
€ 329.64
10g
Please to purchase this item. Need a web account? Register with us today!
11443780
View Documents
Thermo Scientific Alfa Aesar
089459.18
50 g
€ 1376.85
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It used as an intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13454-89-2
413.35
LLVVIWYEOKVOFV-UHFFFAOYSA-L
9801829
[Cu++].[O-][I](=O)=O.[O-][I](=O)=O
CuI2O6
MFCD00016055
copper iodate, copper 2+ diiodate, cupric iodate, copper iodate, anhydrous, cu.2io3
copper;diiodate

Specifications

(decomposition)
Green
95%
MFCD00016055
Soluble in water (1.36g/L).
[Cu++].[O-][I](=O)=O.[O-][I](=O)=O
413.35
413.35
Green powder
Copper iodate, anhydrous
13454-89-2
Powder
CuI2O6
copper iodate, copper 2+ diiodate, cupric iodate, copper iodate, anhydrous, cu.2io3
LLVVIWYEOKVOFV-UHFFFAOYSA-L
copper;diiodate
9801829
Hygroscopic
5.241^1^5
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

missing translation for 'dotInformation' : DOT Class: 5.1, Packing Group: II

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only