missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) oxide, 99.7% (metals basis), Thermo Scientific Chemicals

€ 76.18 - € 767.82

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
Synonym cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
PubChem CID 14829
IUPAC Name copper(2+) oxidandiide
SMILES [O--].[Cu++]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11318838
View Documents
Thermo Scientific Alfa Aesar
041692.22
100 g
€ 76.18
100g
Please to purchase this item. Need a web account? Register with us today!
11338838
View Documents
Thermo Scientific Alfa Aesar
041692.36
500 g
€ 259.29
500g
Please to purchase this item. Need a web account? Register with us today!
11328838
View Documents
Thermo Scientific Alfa Aesar
041692.A3
2 kg
€ 767.82
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Copper(II) oxide is used as an adsorbent and absorbent, pigment, antifouling agent, wood preservative, optical glass polishing agent, welding flux for bronze materials, agricultural chemical and abrasive. It is used in the preparation of intermediates, oxidizing/reducing agents, surface treating agents and process regulators. It finds use in the manufacture of rayons, phosphors, and dry battery cells. It serves as a catalyst in certain chemical reactions of industrial importance. It is also employed in chemical sensors and superconducting materials. Hazardous materials like cyanides, dioxins and halogenated hydrocarbons can be disposed through oxidation with copper oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1317-38-0
79.55
KKCXRELNMOYFLS-UHFFFAOYSA-N
14829
[O--].[Cu++]
CuO
MFCD00010979
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
copper(2+) oxidandiide

Specifications

1326°C
Black
99.7% (Metal basis)
MFCD00010979
Practically insoluble in water and alcohol. Soluble in dilute acids,alkali cyanides,and ammonium carbonate solution.
[O--].[Cu++]
79.55
79.55
2.63
6.315 g/cm3
1317-38-0
Powder
CuO
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
KKCXRELNMOYFLS-UHFFFAOYSA-N
copper(2+) oxidandiide
14829
Odorless
Black powder
Copper(II) oxide
Videos
Safety and Handling

Safety and Handling

H400-H410-H302: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed.

missing translation for 'dotInformation' : DOT Class: 9, Packing Group: III

missing translation for 'einecsNumber' : 215-269-1

missing translation for 'rtecsNumber' : GL7900000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Copper(II) oxide

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Copper oxide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only