Learn More
Copper foil, 1.27mm (0.05in) thick, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11306708
Copper foil, 1.27mm (0.05in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Product Code. 11306708
20 x 100 cm
Click to view available options
Quantity:
20 x 20 cm
20 x 100 cm
Product Code. 11306708
Brand: Thermo Scientific Alfa Aesar 013377.CV
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]
Specifications
Quantity 20 x 100 cm
Percent Purity 99.9%
Odor Odorless
Weight ≈455g/20x20cm
Form Foil
Assay (metals basis)
Chemical Name or Material Copper foil, 1.27mm (0.05 in.) thick
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberL16M50 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number013377.CV,013377.CU
Lot NumberE23T007 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number013377.CV,013377.CU
Lot NumberY07E013 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013377.CV,013377.CU
Lot NumberC22Z037 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013377.CV,013377.CU
Lot NumberL19N25 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013377.CV,013377.CU
RUO – Research Use Only
Product Content Correction
Product Title
Copper foil, 1.27mm (0.05in) thick, 99.9% (metals basis), Thermo Scientific Chemicals >