Learn More
Corning™ CoolRack™ XT PCR Tube Modules
BIOPHARMA
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
Brand:  Corning™ 432055
410.92 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 15502831
  • € 420.00 / Each
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Description
Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to > 100°C and it will rapidly adapt to that temperature.
  • CoolRack modules ensure ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • SBS-compatible.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Capacity: 384-well PCR microplate, SBS-compatible
PCR384
PCR Plate Module
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number04216022 Certificate TypeCertificate of Quality Date30/03/2025 Catalog Number432055
Lot Number04216022 Certificate TypeCertificate of Compliance Date16/05/2023 Catalog Number432055
Lot Number04216022 Certificate TypeCertificate of Analysis Date04/06/2022 Catalog Number432055
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ CoolRack™ XT PCR Tube Modules > CoolRack XT PCR384; 384-well PCR microplate, SBS-compatible