Learn More
Fisherbrand™ CoolCube Microtube and PCR Tube Cooler
Product Code. 16321338 Shop All Fisherbrand Products
Click to view available options
:
Dimensions (L x W x H): 14.3 x 14.3 x 6.4 cm (5.6 x 5.6 x 2.5 in.)
Product Code. 16321338
Only null left
Add to basket
Edge
Add to basket
Protects enzymes and other temperature-sensitive samples from degradation, while working on the laboratory bench. Microtube and PCR Tube Cooler activates overnight in freezer and provides temporary cold storage by eliminating the need for ice buckets.
  • Ideal for temporarily storing temperature-sensitive samples while setting up reactions
  • Maintains 0°C for 4 hours
  • Dual-sided
  • Holds up to 36 x 1.5/2.0 mL microtubes
  • Other side can hold 0.2 mL PCR tubes and strips or a 96-well PCR plate with additional capacity for 10 x 1.5/2.0 mL microtubes
Specifications
Dimensions (L x W x H) 14.3 x 14.3 x 6.4 cm (5.6 x 5.6 x 2.5 in.)
For Use With (Application) Keeping samples cold on the bench without an ice bucket
Weight (English) 2 lb.
Weight (Metric) 1 kg
Holds Temp., Hours 0°C for 4 hours
Color Light Gray
Autoclavable Not Autoclavable