Learn More
Contec™ VertiKlean™ Sox Mop Head Covers
Shop All Contec Products
Click to view available options
Size:
Large
Medium
Regular
For Use With (Equipment):
Large VertiKlean Mop Head
Medium VertiKlean Mop Head
Regular VertiKlean Mop Head
Unit Size:
Pack of 120
Pack of 32
Description
- Clean-processed polyester
- Extends the life of VertiKlean mop heads
Specifications
Specifications
| Product Type | Mop Cover |
| Color Code | White |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Size | Large |
| Disposable | Single-use |
| For Use With (Equipment) | Large VertiKlean Mop Head |
| For Use With (Application) | Removable cover increases durability of VertiKlean mop when used on abrasive surfaces |
| Color-coded | No |
| Material | Polyester |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido