Learn More
WTW™ ConOx Ox-Modul Dissolved Oxygen Sensor Module
Brand:  WTW™ 401070
Product Code. 10044023
Specifications
1 to 2 μS/cm
ConOx with Instruction Manual
Dissolved Oxygen Sensor Module
3 Exchange membrane heads, cleaning solution, abrasive foil, OxiCal CX calibration vessel, electrolyte and beaker
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
WTW™ ConOx Ox-Modul Dissolved Oxygen Sensor Module > Conductivity: 1 to 2�S/cm