Learn More
SCAT Y-Connector
Shop All SCAT Europe Products
Click to view available options
Diameter (Metric):
11 mm
3 mm
4 mm
5 mm
6 to 7 mm
9 mm
Unit Size:
Each
Description
SCAT™ connectors are available in the following versions:
- Conical connectors
- Y-connectors
- Tube connectors
- Capillary connectors
Specifications
Specifications
| Diameter (Metric) | 4 mm |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| For Use With | Tubing |
| For Use With (Application) | HPLC/UHPLC Systems |
| Color | Transparent |
| Product Type | Y Connector |