Learn More
SCAT™ Conical Connector
Product Code. 10327779 Shop All SCAT Europe Products
Click to view available options
Diameter (Metric):
3 to 5 mm
4 to 8 mm
4 to 8 mm, 12 to 16 mm
4 to 8 mm, 8 to 12 mm
5 to 16 mm, 7.5 to 16 mm
7 to 10 mm
8 to 12 mm, 12 to 16 mm
Unit Size:
Each
Product Code. 10327779
Brand: SCAT™ 107825
Only null left
Add to basket
Add to basket
Safe connection of tubes of different sizes: directly and without technical aids. For safe collection of liquid HPLC waste, and many other applications in the laboratory!
SCAT™ connectors are available in the following versions:
  • Conical connectors
  • Y-connectors
  • Tube connectors
  • Capillary connectors
Specifications
Diameter (Metric) 3 to 5 mm
Autoclavable Autoclavable
Material Polypropylene
For Use With Tubing
For Use With (Application) HPLC/UHPLC Systems
Color White
Product Type Conical Connector