Learn More
SCAT Capillary Connector
Shop All SCAT Europe Products
Click to view available options
Diameter (Metric):
1.6 mm, 2.3 mm, 3.2 mm
Unit Size:
Each
Description
SCAT™ connectors are available in the following versions:
- Conical connectors
- Y-connectors
- Tube connectors
- Capillary connectors
Specifications
Specifications
| Diameter (Metric) | 1.6 mm, 2.3 mm, 3.2 mm |
| Autoclavable | Autoclavable |
| Material | PTFE |
| For Use With | Standard Capillaries |
| For Use With (Application) | HPLC/UHPLC Systems |
| Color | White |
| Product Type | Capillary Connector |