Learn More
WTW™ Connection Cable
Brand: WTW™ 903859
938.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Specifications
MPP IDS Respectively IDS Sensors | |
Connection Cable | |
100 m |
Product Content Correction
Product Title
WTW™ Connection Cable > Length: 100m