Learn More
Conductivity Standard 12880μS/cm, With Certificate, Hanna Instruments™
Description
Premium quality 1413 and 12880 μS/cm conductivity solutions produced for analytical use in accordance with ISO 3696/BS3978 standards. They have been standardized with a conductivity meter that was calibrated with potassium chloride NIST 2202 SRM (Standard Reference Material) in deionized water. Additional equipment used in preparation of the solutions include certified weight-checked balances, Class A glassware and certified thermometers.
Buffers are traceable to NIST and are supplied with a Certificate of Analysis that lists the product code, lot number, mean lot value, SRMs that were used, manufacturing date and expiration date.
Traceability and technical information printed on each sachet
- Marked with lot number and expiration date for unopened sachet
- Temperature chart of the actual μS/cm value at various temperatures printed on each sachet
Convenience of individual packaging for one time use
- Ideal for users that do not calibrate frequently
- Ideal for field use due to portability
- Every sachet is as fresh as the day it was packaged
- Light block packaging prevents oxidation from UV light that could alter the buffer value
Specifications
Specifications
Packaging | Sachets |
Quantity | 25 x 20 mL |
Color | Colorless |
Odor | Odorless |
pH | 7 |
Solubility | Soluble |
Physical Form | Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.