missing translation for 'onlineSavingsMsg'
Learn More

12880μS/cm Conductivity Standard Solutions, Hanna Instruments™

Product Code. 10099050
Click to view available options
Quantity:
500 mL
Packaging:
Bottles
This item is not returnable. View return policy
This item is not returnable. View return policy

Premium quality 12880μS/cm conductivity standard that is prepared against a NIST traceable potassium chloride solution

  • Premium quality 12880μS/cm conductivity standard that is prepared against a NIST traceable potassium chloride solution
  • Temperature chart of the actual μS/cm value at various temperatures printed on each bottle
  • Air tight bottle with tamper-proof seal to ensure quality
  • For laboratory and industrial use 
  • 12880μS/cm conductivity solution standardized with a conductivity meter that has been calibrated with potassium chloride NIST 2202 SRM (Standard Reference Material) in deionized water for analytical use in accordance with ISO 3696/BS3978
  • Additional equipment used in preparation of the solutions include certified weight-checked balances, Class A glassware and certified thermometers

Specifications

Packaging Bottles
Quantity 500 mL
Color Colorless
Odor Odorless
pH 7
Solubility Soluble
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.