Learn More
Conductivity Standard 111800μS/cm, Hanna Instruments™
Click to view available options
Quantity:
230 mL
Packaging:
Bottles
Specifications
Specifications
Packaging | Bottles |
Quantity | 230 mL |
Color | Colorless |
Odor | Odorless |
pH | 7 |
Solubility | Soluble |
Physical Form | Liquid |