Learn More
Conductivity Solution Standard, Hanna Instruments
Product Code. 10462521
Click to view available options
Quantity:
460 mL
Unit Size:
460mL
Product Code. 10462521
missing translation for 'mfr': Hanna Instruments undefined
A storage solution prepared with reagent grade chemicals that can be used to ensure optimum performance of your pH and ORP electrodes.
  • Storage solution prepared with reagent grade chemicals that can be used to ensure optimum performance of your pH and ORP electrodes
  • Special formulation to minimize microbial growth and osmotic/diffusion effects between the solution and inner reference electrolyte
  • Manufactured specifically for pH and ORP electrodes
  • Clearly marked expiration date and lot number
Specifications
Chemical Name or Material Conductivity Solution Standard
Quantity 460 mL