Learn More
Lenz Laborglasintrumente Condensers Acc. to Liebig with Olives
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05210402
This item is not returnable.
View return policy
Description
- Liebig condenser acc. to DIN 12576 and in addition to DIN
- For condensing vapours
- With NS socket and NS cone
- Water connection: glass olives
- Made of DURAN™ tubing
Specifications
400 mm | |
NS 19/26 | |
Borosilicate Glass (3.3) |
Product Content Correction
Product Title
Lenz Laborglasintrumente Condensers Acc. to Liebig with Olives > 400 mm