Learn More
Memmert™ Compressed Air Dehumidification For Ich750
For Use With: Constant Climate Chamber
Brand:  Memmert™ ICH750C9
Product Code. 16396687
  • € 2700.00 / Each
Estimated Shipment: 25-06-2025
to see stock.
Explore more special offers
Add to basket
Includes: Standard works calibration certificate
Description
Description
  • Compressed air dehumidification (efficient dehumidification of the interior by means of compressed air)
  • Standard works calibration certificate (measuring point chamber centre) at +10°C with 10 % rh
Specifications
Specifications
Constant Climate Chamber
Compressed Air Dehumidification
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Compressed Air Dehumidification For Ich750 >