Learn More
Agilent BioTek Complete Dispense and Waste Systems
Complete Dispense and Waste Systems include waste and dispense accessories, including racks and tubing, and are for use with various models of microplate washers
Brand:  Agilent BioTek 4070078
 View more versions of this product
Product Code. 16522192
  • € 795.00 / Each
Explore more special offers
Description
Description
4070077 includes: 3 Supply Bottles with Filter, 4 L; 1 Waste Bottle, 4 L; 2 Racks, and Tubing, 4070078 includes: 2 Dispense Bottles with Filter, 4L; 1 Waste Bottle, 4L ; 2 Racks, and Tubing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Waste System
50 TS Microplate Washer, Agilent BioTek Microplate Washer and Reader Instruments
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers