Learn More
Lotz Lagertechnik™ Comfort Swivel Chair
Click to view available options
Adjustable (Height):
435 to 625 mm
465 to 655 mm
480 to 670 mm
525 to 710 mm
Foot Type:
Skid
Wheels
Unit Size:
Each
Specifications
Specifications
| Color | Black |
| Adjustable (Height) | 465 to 655 mm |
| Foot Type | Wheels |
| Upholstery | None |