Learn More
R&D Systems™ CometAssay Lysis Solution (2 x 500 mL)
For use with Single Cell Gel Electrophoresis Assay
Brand:  R&D Systems™ 4250-050-01
Product Code. 30002113
  • € 174.00 / 500mL
Explore more special offers
Description
Description
Cells are lysed using Lysis solution. Cool to 4°C, or on ice, for at least 20 minutes before use.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 x 500 mL
CometAssay Lysis Solution
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems™ CometAssay Lysis Solution (2 x 500 mL) > 2 x 500mL