Learn More
Hoefer™ Combs for HE 33 Mini Submarine Unit
Click to view available options
No. of Wells:
1 Prep, 2 Reference
12
16
8
Preparative
Thickness (Metric):
0.1 cm
0.15 cm
Type:
Backless Comb
Backless Preparative Comb
Comb Back and Two Screws
Unit Size:
Each
Description
- Variety of eight sizes ranging from 1/2 to 16 wells
- 1 to 1.5 mm Thickness
- 44/6 to 2.6 mm Well Width
- 235/35 to 20.5 μL Sample Volume
Specifications
Specifications
| For Use With (Equipment) | HE33 Mini Submarine Unit |
| Type | Backless Comb |
| No. of Wells | 16 |
| Thickness (English) | 0.05 in. |
| Thickness (Metric) | 0.15 cm |
| Volume (Fill) Metric | 20.5 μL |
| Width (English) Well | 0.1 in. |
| Width (Metric) Well | 0.26 cm |
| Depth (English) Pan | 0.19 in. |
| Depth (Metric) Pan | 0.5 cm |