Learn More
Hoefer™ Combs for HE 33 Mini Submarine Unit
Accessory parts for HE 33 Mini Submarine
Brand: Hoefer™ HE31-BK
Description
- Variety of eight sizes ranging from 1/2 to 16 wells
- 1 to 1.5 mm Thickness
- 44/6 to 2.6 mm Well Width
- 235/35 to 20.5 μL Sample Volume
Specifications
HE33 Mini Submarine Unit | |
Comb Back and Two Screws | |
1 Prep, 2 Reference |