Learn More
Hoefer™ Combs for HE 33 Mini Submarine Unit
Accessory parts for HE 33 Mini Submarine
Brand:  Hoefer™ HE31A-P-1.0
 View more versions of this product
Product Code. 11559533
  • € 74.65 / Each
Estimated Shipment: 22-07-2025
to see stock.
Explore more special offers
Description
  • Variety of eight sizes ranging from 1/2 to 16 wells
  • 1 to 1.5 mm Thickness
  • 44/6 to 2.6 mm Well Width
  • 235/35 to 20.5 μL Sample Volume
Specifications
Specifications
HE33 Mini Submarine Unit
Preparative
0.1 cm
1.73/0.2 in.
0.19 in.
Backless Preparative Comb
0.03 in.
235/35 μL
4.4/0.6 cm
0.5 cm
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hoefer™ Combs for HE 33 Mini Submarine Unit > Comb, preparative, backless, 1.0mm thick