Learn More
B Braun™ Combi-Stopper Closing Cones
B Braun™ Combi-Stopper Closing Cones are suitable for sterile closings of IV and syringe assemblies
Brand:  B Braun™ 4495209
 View more versions of this product
Product Code. 10520053
  • € 29.00 / Pack of 100
Edge
Explore more special offers
Description
Description
  • Luer Lock fitting male and female
  • Made of PE (Polyethylene)
  • Sterile closing of all kinds of open IV-accesses in IV-Sets
  • Sterile closing of pre-filled syringes
  • Available in red, blue and white
  • Box of 100 pcs
Specifications
Specifications
Polyethylene
Combi Stopper
Sterile
White
Luer Lock
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
B Braun™ Combi-Stopper Closing Cones > Color: White