Learn More
Lenz Laborglasinstrumente™ Column Head with Valve Stopcock
Made of DURAN™ tubing. Lenz Laborglasinstrumente™ Column Head with Valve Stopcock has a PTFE for easy and accurate adjustment and ground joint NS 14/23 for thermometers with a fitting length of 55 mm.
Brand: Lenz Laborglasinstrumente™ 05170111
This item is not returnable.
View return policy
Description
- With PTFE valve
- Made of DURAN™ tubing
Specifications
NS 14/23 | |
DURAN Tubing | |
PTFE Valve | |
1 Each |
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Column Head with Valve Stopcock >