Learn More
Thermo Scientific™ Columbia Agar with Sheep Blood PLUS/MacConkey Agar without Salt Biplate
Isolate and cultivate fastidious microorganisms while simultaneously detecting and Gram-negative pathogens from urine samples using this bi-plate.
Brand: Thermo Scientific™ PB5254E
17.12 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Isolate and cultivate fastidious microorganisms with clearly visible hemolytic reactions (staphylococci and streptococci), while simultaneously detecting and Gram-negative pathogens from urine samples using Thermo Scientific™ Oxoid™ Columbia Agar with Sheep Blood PLUS/MacConkey Agar without Salt Biplate.
The biplate format combines a selective and a general purpose medium on one plate reducing the need for further confirmation steps by allowing the laboratories to isolate pathogens directly without any subculture.
Produce more diagnostic details of pathogenic organisms while reducing storage capacity issue in the incubator with a bi-plate that combines Gram-positive and Gram-negative organism detection on a single plate, allowing the laboratories get the full clinical picture for the next diagnostic decision.
- Reduced confirmatory testing: combines selective and a general medium on a single plate, allowing the laboratories to isolate the pathogens directly without any subculture for further confirmation steps.
- Prevents the overgrowth of non-target organisms: MacConkey Agar without salt provides a low electrolyte medium on which most Proteus species do not spread making it suitable for examination of urine samples
- Suitable for identification of organism occurring in urinary tract infections.
- Ready-to-use convenience of prepared media
- Space-saving bi-plate format
Columbia Blood Agar with Sheep Blood PLUS promotes rapid production of large colonies, clearly defined zones of hemolysis and good colonial differentiation while MacConkey Agar without Salt suppresses the swarming of Proteus species and is therefore recommended for urine examination in the investigation of suspected urinary tract infection (UTI).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
Red, opaque/ Salmon pink, transparent | |
Columbia Agar with Sheep Blood PLUS/ MacConkey Agar without Salt Biplate | |
United Kingdom | |
17g ± 5% | |
10 Pack |
6°C to 12°C | |
Biplate | |
Agar Plate | |
Flow Wrapped - Single Layer |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
4497408 | Certificate of Analysis | 14/03/2025 | PB5254E |
4495858 | Certificate of Analysis | 07/03/2025 | PB5254E |
4494900 | Certificate of Analysis | 03/03/2025 | PB5254E |
4492461 | Certificate of Analysis | 23/02/2025 | PB5254E |
4491750 | Certificate of Analysis | 18/02/2025 | PB5254E |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.