Learn More
Zeiss™ Cold Light Source
Product Code. 15394693
Click to view available options
Type:
CL 1500 HAL
KL 2500 LCD
Unit Size:
Each
Product Code. 15394693
Brand: Zeiss™ 4357109000000
Only null left
Add to basket
Add to basket
Designed for the most challenging applications in Stereo Microscopy, Macroscopy and Forensics
Cold-light Source KL 2500 LCD
  • Supplied with a filter wheel, which accommodates up to five insert filters
  • Filter selection is as simple as a turn of the knob
  • Electronic accessories (remote control, RS-232 or tilt switch), connect to the front panel of the device enabling light source operation in difficult access areas.
  • Modified light guide socket
  • Light guides up to 15mm fiber bundle diameter can be used with optimum results
  • Design compatibility: KL 1500 LCD light guides can be used with the KL 2500 LCD
  • All other functions are identical to the KL 1500 LCD
Compatible Microscopes:
  • Axio Zoom.V16
  • Axioskop 2 FS MOT
  • Axioskop 2 FS Plus
  • Stemi 2000
  • Stemi 2000 C
  • Stemi 2000 CS
  • STEMI SV 11
  • STEMI SV 6
  • SteREO Discovery.V12
  • SteREO Discovery.V20
  • SteREO Lumar.V12
Cold-light source CL 6000 LED (D)
  • Up to 600 lm light flux at 9 mm fiber output dia., ca. 6200K color temperature
  • Min. 50,000 operating hours until intensity drop to 70%
  • Control of intensity and 6 memory cells by rotary push knob and LCD
  • 10 mm collet chuck with sensor for 1500 light guides
  • Filter slider for 2 mounted filters 35x26x4 mm and with free opening
  • Max. 70W wide-range supply for flicker free light, silent fan
  • 2.5 mm jack bush for on/off via optional foot button S, USB socket
  • Country-specific mains cord
  • Dimensions D241xW192xH118 mm, weight 3.1 kg
Compatible Microscopes:
  • Axio Zoom.V16
  • Stemi 2000
  • Stemi 2000 C
  • Stemi 2000 CS
  • Stemi DV4
  • SteREO Discovery.V12
  • SteREO Discovery.V20
  • SteREO Discovery.V8
Cold light source CL 1500 HAL
  • Up to 600lm light flux at 9mm optical output dia., continuous electronically dimming,
  • LCD display for brightness in % or color temperature in K or power-on hours,
  • Incl. bulb 15V 150W halogen reflector, lifetime ca. 150h at 80%=450lm,
  • 10mm clamping nut with sensor for light guide 1500,
  • Filter slider for one filter d=28mm and with free opening,
  • Max. 180W wide-range supply for flicker free light, silent axial fan,
  • Country-specific mains cord
Compatible Microscopes:
  • Axio Zoom.V16
  • Stemi 2000
  • Stemi 2000 C
  • Stemi 2000 CS
  • Stemi DV4
  • SteREO Discovery.V12
  • SteREO Discovery.V20
  • SteREO Discovery.V8
Specifications
Type CL 1500 HAL
For Use With (Application) Stereo Microscopy, Macroscopy and Forensics
For Use With (Equipment) Carl Zeiss Microscopes
Quantity 1 Ea.