Learn More
Lenz Laborglasinstrumente™ Coil Springs
Click to view available options
Overall Length (Metric):
60 mm
Unit Size:
Pack of 10
Description
- Made from stainless steel
- Two eyelets
Specifications
Specifications
| Overall Length (Metric) | 60 mm |
| Packaging Quantity | 10 |