Learn More
Hach Lange™ COD Cuvette Test
Brand:  Hach Lange™ LCK1014
Product Code. 15117236
  • € 164.32 / Pack of 25
Explore more special offers
Specifications
ISO 6060-1989, DIN 38409-H41-H44
25
Dichromate
For Automatic Recognition in the Photometer
100 to 2000mg/L O2
COD Cuvette Test
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hach Lange™ COD Cuvette Test > Range: 100 to 2000mg/L O2