missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt for AAS 1g/L HCl matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
€ 25.95 - € 76.50
Chemical Identifiers
CAS | 7697-37-2 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12963135
|
Fisher Chemical
B 012 100 |
100 mL |
€ 25.95
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12983135
|
Fisher Chemical
B 012 500 |
500 mL |
€ 47.55
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12973135
|
Fisher Chemical
B 012 1000 |
1000 mL |
€ 76.50
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
CHEBI:48107 | |
O[N+]([O-])=O |
HNO3 | |
MFCD00011349 | |
944 | |
nitric acid |
Specifications
1g/L ±0.002g/L | |
Bottle | |
HNO3 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
2% HCl | |
Cobalt |
7697-37-2 | |
Flame Single-element Standard Solution | |
MFCD00011349 | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 | |
Liquid |
Safety and Handling
H350 ; H412
P201;P202;P273;P308+P313;P405;P280;P501
Danger
missing translation for 'shelfLife' : 24 Months. (12 Months after opening)
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Single-element Standard Solution for AAS. Cobalt (Co) 1000mg/l in HCl 2%
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H350-May cause cancer.
- H412-Harmful to aquatic life with long lasting effects.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P273-Avoid release to the environment.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P501-Dispose of contents/container to .