missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Clyde S3 Shoe

If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Clyde.

€ 136.00 - € 146.00

Specifications

Certifications/Compliance ISO 20345:2011
Closure Type Laces
Color Black
For Use With (Application) Logistics and Transport, Electronics, Construction and Technology, Manufacturing, Nature and Environment
Liner Material Hydro-Tec Antibacterial
View More Specs

Products 10
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17532666
View Documents
Emma Safety Footwear
MM99656939
39
€ 136.00
1 pair
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
 
17542666
View Documents
Emma Safety Footwear
MM99656940
40
€ 145.00
1 pair
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
 
17552666
View Documents
Emma Safety Footwear
MM99656941
41
€ 136.00
1 pair
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
 
17562666
View Documents
Emma Safety Footwear
MM99656942
42
€ 146.00
1 pair
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
 
17572666
View Documents
Emma Safety Footwear
MM99656943
43
€ 136.00
1 pair
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
 
17582666
View Documents
Emma Safety Footwear
MM99656944
44
€ 136.00
1 pair
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
 
17592666
View Documents
Emma Safety Footwear
MM99656945
45
€ 136.00
1 pair
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
 
17502676
View Documents
Emma Safety Footwear
MM99656946
46
€ 136.00
1 pair
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
 
17512676
View Documents
Emma Safety Footwear
MM99656947
47
€ 136.00
1 pair
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
 
17522676
View Documents
Emma Safety Footwear
MM99656948
48
€ 136.00
1 pair
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
 
Description

Description

  • S3-Safetyshoe with sole plate for wet work
  • Metal Free
  • The streetborn safety shoe. If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Clyde.
  • A no compromise metal free safety shoe that looks and feels like a classic sneaker.
  • It gets its name from the highly supportive reinforced heel. This heel together with the shoe's reinforced, shock absorbing sole keeps ankles, shins and joints free from stress and injury.
  • DenimX laces with a story. Made from recycled jeans for our circular safety shoes.
  • Industries:
    • Logistics & transport
    • Electronics
    • Construction & technology
    • Manufacturing
    • Nature & environment
Specifications

Specifications

ISO 20345:2011
Black
Hydro-Tec Antibacterial
Extra-wide Width
Safety Shoe
Laces
Logistics and Transport, Electronics, Construction and Technology, Manufacturing, Nature and Environment
Nubuck, Oiled Nubuck
High
Videos
SDS
Documents

Documents

Product Certifications