missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris™ CLP 290
K+/Cl- cotransporter 2 (KCC2) activator; orally bioavailable prodrug of CLP 257 (Cat. No.5242)
€ 275.37 - € 1078.81
Specifications
Chemical Name or Material | (Z)-5-Fluoro-2-((4-oxo-2-(tetrahydropyridazin-1(2 H)-yl)thiazol-5(4 H)-ylidene)methyl)phenyl pyrrolidine-1-carboxylate |
---|---|
CAS | 1181083-81-7 |
Target | KCC2 Activators |
Molecular Formula | C19H21FN4O3S |
Purity | 98% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18766866
|
Tocris™
6758/10 |
10 mg |
€ 275.37
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18787986
|
Tocris™
6758/50 |
50 mg |
€ 1078.81
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
K+/Cl- Cotransporter 2 (KCC2) activator. Activates spinal inhibitory interneurons, restores physiological neuronal activity in the spinal cord and induces functional recovery in a mouse model of spinal cord injury. Also decreases blood vasopressin and glucose levels in mice with Streptozotocin (Cat. No. 1621) induced diabetes mellitus. Orally bioavailable prodrug of CLP 257 (Cat. No.5242).Specifications
(Z)-5-Fluoro-2-((4-oxo-2-(tetrahydropyridazin-1(2 H)-yl)thiazol-5(4 H)-ylidene)methyl)phenyl pyrrolidine-1-carboxylate | |
KCC2 Activators | |
98% |
1181083-81-7 | |
C19H21FN4O3S |
Safety and Handling
Recommended Storage : Store at -20°C