Learn More
Tocris Bioscience™ CLP 257
Selective KCC2 activator
Brand:  Tocris Bioscience™ 5242/50
Additional Details : CAS : 1181081-71-9 Weight : 0.00970kg
 View more versions of this product
Product Code. 18797199
  • € 1034.53 / 50mg
Estimated Shipment: 08-07-2024
to see stock.
Description
Description
CLP 257 is a selective KCC2 K+-Cl- cotransporter activator (EC50 = 616 nM). Exhibits selectivity for KCC2 over other KCC family members, NKCC1 and GABAA receptors, as well as 55 other receptors. Enhances Cl- transport and increases KCC2 plasma membrane expression in spinal slices with reduced KCC2 function. Alleviates hypersensitivity in rats with peripheral nerve injury.
Specifications
Specifications
(5Z)-5-[(4-Fluoro-2-hydroxyphenyl)methylene]-2-(tetrahydro-1-(2H)-pyridazinyl)-4(5H)-thiazolone
50 mg
C14H14FN3O2S
>98%
1181081-71-9
KCC2 Activators
307.34
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers