missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Verenigde Kunststof Bedrijven™ HDPE, Alveolit Sealring, Screw Closure DIN 61 (Gray)
Click to view available options
Color:
Gray
Description:
Screwcap DIN 61
Description
- HDPE screw cap
- leak-proof closure
- Applicable on plastic packages
Specifications
Specifications
Color | Gray |
Description | Screwcap DIN 61 |
For Use With (Equipment) | Bottle and Jerrican |
Material | HDPE, Alveolit |
Product Type | Screw Closure |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction