Learn More
Thermo Scientific™ CLINIConic Fixed-Angle Rotor for Compact and 1.6 L General Purpose X Pro and ST Plus Centrifuges
Shop All Thermo Scientific Products
Click to view available options
Capacity:
30 x 15 mL
Unit Size:
Each
Description
Optimize separations with fast acceleration of 30 blood tubes or 15mL conicals in a fixed angle configuration with the Thermo Scientific™ CLINIConic™ Fixed Angle Rotor.
Spin 30 blood tubes or 15mL conicals in a fixed angle configuration with fast acceleration to optimize applications.
Ordering Information:
Tube adapters are sold separately; order rotor body (75-003-623) and components to complete assembly. Contact your sales representative for information.
Specifications
Specifications
| Description | CLINIConic Fixed-Angle Rotor |
| Type | Fixed Angle Rotor |
| Capacity | 30 x 15 mL |
| Max. RCF | 4,997 x g |
| Max. Speed | 5,650 rpm |
| Radius (Metric) | 140 mm |
| Max. Tube Length | 131 mm |
| Max. Tube Diameter | 16.5 mm |
| For Use With (Equipment) | Thermo Scientific Sorvall Legend X1, Sorvall ST 16, Sorvall ST 8, Multifuge X1, Megafuge 16, Megafuge 8, SL 16 and SL 8 Centrifuges |
| Angle | 37° |