Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Straight Reducer, Natural Kynar™
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Brand:  Cole-Parmer 50108-91
 View more versions of this product
Product Code. 30054377
  • € 28.85 / Pack of 10
Estimated Shipment: 04-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. Biopharma compatible and USP 661 and BPOG compliant
Specifications
Specifications
USP;Non-DEHP;Non-Phthalate
10/Pk.
Hose Barb Reducer
Straight
-35°C
1/4 to 1/16 in.
E-beam/gamma irradiation, EtO, or autoclave up to 135°C
Natural
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer Cleanroom Hose Barb Fittings, Straight Reducer, Natural Kynar™ > 1/4 to 1/16 in.