Learn More
Sampling Systems™ Cleaning Coupons for Plastics
Test and validate your cleaning regime with this range of cleaning validation coupons for various plastics.
Brand:  Sampling Systems™ 8644PVC-50
 View more versions of this product
Product Code. 15869551
  • € 33.85 / Each
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Description
  • Tests available for HDPE, nylon, PP, PTFE and PVC
  • Two sizes available for each configuration, either 50 x 50mm or 100 x 100mm
Specifications
Specifications
Cleaning Coupon
50 x 50 mm
Testing and validating cleaning regime
PVC
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sampling Systems™ Cleaning Coupons for Plastics > Material: PVC; Size: 50 x 50mm