Learn More
Sampling Systems™ Cleaning Coupons for Plastics
Product Code. 15829551 Shop All Sampling Systems Products
Click to view available options
Material:
HDPE
Nylon
PTFE
PVC
Polypropylene
Dimensions (L x W):
100 x 100 mm
50 x 50 mm
Unit Size:
Each
Product Code. 15829551
Brand: Sampling Systems™ 8644N50
Only null left
Add to basket
Add to basket
Test and validate your cleaning regime with this range of cleaning validation coupons for various plastics.
  • Tests available for HDPE, nylon, PP, PTFE and PVC
  • Two sizes available for each configuration, either 50 x 50mm or 100 x 100mm
Specifications
Product Type Cleaning Coupon
Dimensions (L x W) 50 x 50 mm
For Use With (Application) Testing and validating cleaning regime
Material Nylon