Learn More
PURUS™ Critical Print Cleanroom Notebooks
Shop All Purus Products
Click to view available options
Color:
Blue
White
Size:
A4
A5
A6
Ruled:
Grid
Yes
Quantity:
10/Cs.
20/Cs.
40/Cs.
Unit Size:
Pack of 10
Pack of 20
Pack of 40
Description
For writing in cleanroom environments
- Cleanroom processed and packaged
- Suitable for use in ISO 4 to 6 (Class 10 to Class 1000) cleanrooms
- Proprietary Binding Technology without organic fillers
- Recyclable
- College Rule
- Amide and Silicone Free
Specifications
Specifications
| Material | Polyethylene |
| Ruled | Grid |
| Color | White |
| Cover Material | Poly |
| Dimensions (L x W) | 148 x 105 mm |
| Width (Metric) | 105 mm |
| Autoclavable | Autoclavable |
| Certifications/Compliance | Suitable for use in ISO 4 to 6 (Class 10 to Class 1000) |
| For Use With (Application) | For writing in cleanroom environments |
| Product Type | Cleanroom Notebook |
| Show More |