Learn More
Huenersdorf™ CLASSIC™ 24-Compartment Assortment Box
These assortment boxes are manufactured of extra dimensionally stable transparent polystyrene with 24 compartments for storage.
Brand: Huenersdorf™ 612500
Description
- Transparent lid and bottom for optimum display of contents
- With stable hinges and closures
- Ideal for packaging purposes
- Food-safe and temperature resistant from −20° to +70°C
Specifications
Hinged lid | |
24 | |
55 mm | |
335 mm |
Transparent | |
225 mm | |
Polystyrene |