Learn More
Witeg™ Class B Burettes
With short line Schellbach blue graduations.
Brand: Witeg™ 3 000 504
This item is not returnable.
View return policy
Description
- With lateral stopcock
- With solid ST glass plug
- Made of clear borosilicate glass 3.3
- DIN ENO ISO 385
Specifications
Clear | |
0.1 mL | |
Glass | |
Glass | |
Buret | |
EN ISO 385 |
Borosilicate Glass | |
Dosing, Titration | |
Lateral | |
±0.08 mL | |
Class B |