Learn More
Witeg™ Class B Burettes
With short line Schellbach blue graduations.
Brand:  Witeg™ 3 000 504
Product Code. 10698305
  • € 43.00 / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With lateral stopcock
  • With solid ST glass plug
  • Made of clear borosilicate glass 3.3
  • DIN ENO ISO 385
Specifications
Specifications
Clear
0.1 mL
Glass
Glass
Buret
EN ISO 385
Borosilicate Glass
Dosing, Titration
Lateral
±0.08 mL
Class B
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers