Learn More
Hirschmann™ Class AS Brown Graduated Zero Point Measuring Pipettes
Provides an extremely high degree of volumetric accuracy for your pipetting needs.
Brand: Hirschmann™ 1100342
This item is not returnable.
View return policy
Description
- With silver diffusion ink for permanent markings
- Brown graduation marks
- Main point ring graduation
- Zero point at bottom
- Manufactured from AR glass™ soda-lime glass with a smooth, non-porous surface
- Certified conformity, with dated batch identification
- Graduated to tip
- Calibrated for EX
- Type 2, DIN EN ISO 835
Specifications
DIN EN ISO 835 | |
AR-GLAS | |
Yes | |
0.1 mL | |
2 mL | |
Measuring Pipet |
AS | |
Volumetric accuracy in pipetting | |
Brown | |
Diffusion ink | |
360 ±5 mm |