Learn More
Hirschmann™ Class AS Brown Graduated Zero Point Measuring Pipettes
Provides an extremely high degree of volumetric accuracy for your pipetting needs.
Brand:  Hirschmann™ 1100365
 View more versions of this product
Product Code. 10426513
  • € 96.60 / Pack of 6
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With silver diffusion ink for permanent markings
  • Brown graduation marks
  • Main point ring graduation
  • Zero point at bottom
  • Manufactured from AR glass™ soda-lime glass with a smooth, non-porous surface
  • Certified conformity, with dated batch identification
  • Graduated to tip
  • Calibrated for EX
  • Type 2, DIN EN ISO 835
Specifications
Specifications
DIN EN ISO 835
AR-GLAS
Yes
0.1 mL
20 mL
Measuring Pipet
AS
Volumetric accuracy in pipetting
Brown
Diffusion ink
360 ±5 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers