Learn More
Hirschmann™ AR™ Glass Volumetric Pipettes, Class AS
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann Instruments™ AR™ Glass Volumetric Pipettes, Class AS come with blue graduation.
Brand:  Hirschmann™ 1341129
 View more versions of this product
Product Code. 10524061
  • € 20.30 / Pack of 6
In Stock
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Acc. to DIN EN ISO 648
  • One mark, blue graduation
  • Conformity certified, with dated batch identification
Specifications
Specifications
± 0.005 mL
To Deliver
AS
Non-sterile
Blue
12/Pack
0.5 mL
Reusable
Autoclavable
DIN EN ISO 648
Soda Lime Glass
Yes
Blue enamel color
± 0.005 mL
Volumetric Pipet
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers