Learn More
Fisherbrand™ Cellulose Stoppers
For flasks, erlenmeyers and test tubes.
Brand:  Fisherbrand™ 0005N00059
 View more versions of this product
Product Code. 11760096
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Permeable in air
  • Sterilizable to 200°C maximum
  • Single use
Specifications
Specifications
Single-use
37 mm
200°C
17 mm
Stopper
Beakers, Erlenmeyer flasks, tubes, and different types of bottles
1000PK
12 mm
Cellulose Fiber
Sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers