missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Vacutainer Citrate Tubes
Click to view available options
Capacity (Metric):
6 mL
Closure Color:
Light Blue
Unit Size:
Pack of 100
Description
- Trisodium citrate is used as an anticoagulant for coagulation investigations.
- It works as an anticoagulant as it forms complexes with metal ions such as calcium inhibiting the coagulation cascade.
- Anticoagulation with trisodium citrate is reversible.
- Blood to additive ratio is 9:1.
- Citrate solution acts as a buffer.
- Suitable for carrying out special test procedures such as the platelet function test PFA100.
- Internal coating of a special silicone to minimize contact activation.
Specifications
Specifications
| Additive | Sodium Citrate (0.105M) |
| Material | Glass |
| Closure Type | Hemogard |
| Capacity (Metric) | 6 mL |
| Closure Color | Light Blue |
| Tube Size | 13 x 100 mm |
| For Use With (Application) | Venous blood sampling, coagulation analysis |
| Label Type | Paper |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction